Abnova
Manufacturer Code:H00008531M06
Catalog # 89-001-086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H5 |
Immunogen | CSDA (NP_003642 241 a.a. ~ 330 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CSDA.
For Research Use Only
Protein Aliases: cold shock domain protein A; Cold shock domain-containing protein A; cold-shock domain containing A1; cold-shock domain protein A; CSDA1; DBPA; DNA-binding protein A; Single-strand DNA-binding protein NF-GMB; Y box binding protein 3; Y-box-binding protein 3; ZO-1-associated nucleic acid-binding protein; ZONAB
Gene Aliases: CSDA; CSDA1; DBPA; YBX3; ZONAB
UniProt ID: (Human) P16989
Entrez Gene ID: (Human) 8531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.