Abnova
Manufacturer Code:H00007812B01
Catalog # 89-006-514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CSDE1 (NP_009089.4 1 a.a. ~ 767 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human CSDE1 protein.
For Research Use Only
Protein Aliases: cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; Cold shock domain-containing protein E1; D1S155E; DKFZp779B0247; DKFZp779J1455; FLJ26882; N-ras upstream gene protein; NRAS-related; Protein UNR; RP5-1000E10.3; UNR; upstream of NRAS
Gene Aliases: CSDE1; D1S155E; KIAA0885; NRU; UNR
UniProt ID: (Human) O75534
Entrez Gene ID: (Human) 7812
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.