Abnova
Manufacturer Code:H00001486D01P
Catalog # 89-017-547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CTBS (NP_004379.1 1 a.a. ~ 385 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human CTBS protein.
For Research Use Only
Protein Aliases: chitobiase, di-N-acetyl-; CTB; Di-N-acetylchitobiase
Gene Aliases: CTB; CTBS
UniProt ID: (Human) Q01459
Entrez Gene ID: (Human) 1486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.