Abnova
Manufacturer Code:H00023705A01
Catalog # 89-004-571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CADM1 (NP_055148 151 a.a. ~ 250 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant CADM1.
For Research Use Only
Protein Aliases: BL2; CADM1; Cell adhesion molecule 1; DKFZp686F1789; IGSF4; IGSF4A; Immunoglobulin superfamily member 4; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4D variant 1; immunoglobulin superfamily, member 4D variant 2; MGC149785; MGC51880; NECL-2; NECL2; nectin-like 2; Nectin-like protein 2; RA175; SgIgSF; Spermatogenic immunoglobulin superfamily; ST17; sTSLC-1; Synaptic cell adhesion molecule; SYNCAM; synCAM1; truncated CADM1 protein; TSLC-1; TSLC1; TSLC1/Nectin-like 2/IGSF4; Tumor suppressor in lung cancer 1
Gene Aliases: BL2; CADM1; IGSF4; IGSF4A; Necl-2; NECL2; RA175; sgIGSF; ST17; sTSLC-1; SYNCAM; synCAM1; TSLC1
UniProt ID: (Human) Q9BY67
Entrez Gene ID: (Human) 23705
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.