Abnova
Manufacturer Code:H00001496B01
Catalog # 89-005-196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CTNNA2 (NP_004380.2 1 a.a. ~ 905 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human CTNNA2 protein.
For Research Use Only
Protein Aliases: Alpha N-catenin; Alpha-catenin-related protein; alpha-N-catenin; cadherin-associated protein, related; cancer/testis antigen 114; CAP-R; CAPR; catenin (cadherin-associated protein), alpha 2; Catenin alpha-2; CTNR; DKFZp686H02198
Gene Aliases: CAP-R; CAPR; CT114; CTNNA2; CTNR
UniProt ID: (Human) B7Z2W7
Entrez Gene ID: (Human) 1496
Molecular Function:
actin family cytoskeletal protein
cell adhesion molecule
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.