Abnova
Manufacturer Code:H00001453A01
Catalog # 89-004-083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CSNK1D (AAH03558 301 a.a. ~ 415 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant CSNK1D.
For Research Use Only
Protein Aliases: casein kinase 1, delta; Casein kinase I isoform delta; CKI-delta; CKId; HCKID; Tau-protein kinase CSNK1D
Gene Aliases: ASPS; CKIdelta; CSNK1D; FASPS2; HCKID
UniProt ID: (Human) P48730
Entrez Gene ID: (Human) 1453
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.