Proteintech
Manufacturer Code:102921AP150UL
Catalog # 50-553-541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | calreticulin Fusion Protein Ag0325 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CABP3; CALBP; Calcium-binding protein 3; CALR Calregulin calreticulin cC1qR CRP55 CRT CRTC ERp60 grp60 HACBP RO SSA; Calregulin; Calreticulin; CRP55; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; grp60; HACBP; Sicca syndrome antigen A (autoantigen Ro; calreticulin)
Gene Aliases: CALR; Calregulin; cC1qR; CRT; CRTC; HEL-S-99n; RO; SSA
UniProt ID: (Human) P27797, (Mouse) P14211, (Rat) P18418
Entrez Gene ID: (Human) 811, (Mouse) 12317, (Rat) 64202
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.