Invitrogen
Manufacturer Code:PA551700
Catalog # PIPA551700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human cIAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIP1 API2 CIAP2 HAIP1 HIAP1 MALT2 MIHC RNF49 c-IAP2 IAP homolog C IAP-1 RING finger protein 49 TNFR2-TRAF signaling complex protein TNFR2-TRAF-signaling complex protein 1 apoptosis inhibitor 2 baculoviral IAP repeat-containing 3 baculoviral IAP repeat-containing protein 3 inhibitor of apoptosis protein 1 mammalian IAP homolog C; API2; apoptosis inhibitor 1; Apoptosis inhibitor 2; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; Baculoviral IAP repeat-containing protein 3; C-IAP2; Cellular inhibitor of apoptosis 2; hIAP-1; IAP homolog C; IAP-1; Inhibitor of apoptosis protein 1; mammalian IAP homolog C; mIAP-1; mIAP1; RING finger protein 49; RING-type E3 ubiquitin transferase BIRC3; TNFR2-TRAF signaling complex protein; TNFR2-TRAF-signaling complex protein 1
Gene Aliases: AIP1; Api1; API2; AW107670; Birc2; BIRC3; c-IAP2; cIAP-1; cIAP-2; cIAP1; CIAP2; HAIP1; HIAP1; HIAP2; IAP1; IAP2; MALT2; MIAP1; MIAP2; MIHB; MIHC; RNF49
UniProt ID: (Human) Q13489, (Mouse) O08863
Entrez Gene ID: (Human) 330, (Mouse) 11796, (Rat) 78971
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.