Invitrogen
Manufacturer Code:MA535343
Catalog # PIMA535343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0168 |
Immunogen | A synthesized peptide derived from human BIRC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: API1 Api2 AW146227 Baculoviral IAP repeat-containing protein 2 Birc2 Birc3 Cellular inhibitor of apoptosis 1 cIAP1 C-IAP1 cIAP2 HIAP1 HIAP2 hIAP-2 IAP homolog B IAP1 IAP2 Inhibitor of apoptosis protein 2 mcIAP1 MIAP1 mIAP2 MIHB MIHC rIAP1 RING finger protein 48 RING-type E3 ubiquitin transferase BIRC2 RNF48 TNFR2-TRAF-signaling complex protein 2; apoptosis inhibitor 1; apoptosis inhibitor 2; baculoviral IAP repeat-containing 2; Baculoviral IAP repeat-containing protein 2; C-IAP1; Cellular inhibitor of apoptosis 1; hIAP-2; IAP homolog B; IAP-2; inhibitor of apoptosis protein; Inhibitor of apoptosis protein 2; NFR2-TRAF signalling complex protein; RING finger protein 48; RING-type E3 ubiquitin transferase BIRC2; TNFR2-TRAF-signaling complex protein 2
Gene Aliases: API1; Api2; BIRC2; c-IAP1; cIAP1; Hiap-2; HIAP2; MIHB; rIAP1; RNF48
UniProt ID: (Human) Q13490
Entrez Gene ID: (Human) 329, (Rat) 60371
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.