R&D Systems
Manufacturer Code:AF8181
Catalog # AF8181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human cIAP-1/HIAP-2 His2-Ser618 Accession # Q13490 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: API1Hiap-2 apoptosis inhibitor 1 baculoviral IAP repeat containing 2 baculoviral IAP repeat-containing 2 baculoviral IAP repeat-containing protein 2 BIRC2 cIAP1 C-IAP1 hIAP2 HIAP-2 IAP homolog B IAP2 IAP-2 Inhibitor of apoptosis protein 2 MIHB MIHBHIAP2 NFR2-TRAF signalling complex protein RING finger protein 48 RNF48hiap-2 TNFR2-TRAF-signaling complex protein 2; apoptosis inhibitor 1; baculoviral IAP repeat-containing 2; Baculoviral IAP repeat-containing protein 2; C-IAP1; Cellular inhibitor of apoptosis 1; hIAP-2; IAP homolog B; IAP-2; Inhibitor of apoptosis protein 2; NFR2-TRAF signalling complex protein; RING finger protein 48; RING-type E3 ubiquitin transferase BIRC2; TNFR2-TRAF-signaling complex protein 2
Gene Aliases: API1; BIRC2; c-IAP1; cIAP1; Hiap-2; HIAP2; MIHB; RNF48
UniProt ID: (Human) Q13490
Entrez Gene ID: (Human) 329
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.