Invitrogen
Manufacturer Code:PA5116863
Catalog # PIPA5116863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human PRKG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW212535 CGK 2 cGK2 cGKII cGK-II cGMP dependent protein kinase type II cGMP-dependent protein kinase 2 cGMP-dependent protein kinase II EC 2.7.11.12 GDPKII Prkg2 PRKGR2 protein kinase cGMP- dependent type 2 protein kinase cGMP- dependent type 2 protein kinase cGMP- dependent type II protein kinase cGMP-dependent type II testicular tissue protein Li 99 type II cGMP-dependent protein kinase; CGK 2; cGK2; cGKII; cGMP dependent protein kinase type II; cGMP-dependent protein kinase 2; cGMP-dependent protein kinase II; protein kinase cGMP- dependent type 2; protein kinase, cGMP- dependent, type 2; protein kinase, cGMP- dependent, type II; testicular tissue protein Li 99; type II cGMP-dependent protein kinase
Gene Aliases: AW212535; cGK-II; cGK2; CGKII; GDPKII; PRKG2; PRKGR2
UniProt ID: (Human) Q13237, (Rat) Q64595
Entrez Gene ID: (Human) 5593, (Rat) 25523, (Mouse) 19092
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.