Invitrogen
Manufacturer Code:PA5102630
Catalog # PIPA5102630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PRKG1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAT8 AW125416 cGK cGK 1 CGK 1 alpha cGK 1 beta cGK cGK cGKI cGK cGK1 cGK1 cGKI cGKI-alpha cGKI-BETA cGMP dependent protein kinase type 1 cGMP kinase type I alpha cGMP-dependent protein kinase 1 cGMP-dependent protein kinase 1 alpha isozyme cGMP-dependent protein kinase 1 beta isozyme cGMP-dependent protein kinase I Gm19690 OTTHUMP00000019618 PKG Pkgi PRKG1 Prkg1b PRKGR1A Prkgr1b protein kinase cGMP-dependent 1 protein kinase cGMP-dependent regulatory type I beta protein kinase cGMP-dependent type 1 protein kinase cGMP-dependent type I RP11-346D6.1 Unknown (protein for MGC:134126) unnamed protein product; cGK 1; cGK1; cGKI; cGMP dependent protein kinase type 1; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase I; protein kinase, cGMP-dependent, regulatory, type I, beta; protein kinase, cGMP-dependent, type I
Gene Aliases: AAT8; AW125416; cGK; cGK 1; cGK1; CGKI; cGKI-alpha; cGKI-BETA; Gm19690; PKG; Pkgi; PRKG1; PRKG1B; PRKGR1A; PRKGR1B
UniProt ID: (Human) Q13976, (Mouse) P0C605
Entrez Gene ID: (Human) 5592, (Mouse) 19091, (Rat) 54286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.