Novus Biologicals
Manufacturer Code:NBP271227R
Catalog # NBP271227R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9G4 |
Immunogen | Full length recombinant protein of human cGK1/PRKG1 (NP_006249) produced in HEK293T cells. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AAT8 cGK cGK 1 cGK1 cGKI cGKI-alpha cGKI-BETA cGMP-dependent protein kinase I DKFZp686K042 EC 2.7.11 EC 2.7.11.12 FLJ36117 PKG PKG1 PRKG1B PRKG1BMGC71944 PRKGR1A PRKGR1B PRKGR1BcGMP-dependent regulatory type I beta protein kinase cGMP-dependent 1 protein kinase cGMP-dependent type I; cGK 1; cGKI; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase I; protein kinase, cGMP-dependent, regulatory, type I, beta
Gene Aliases: AAT8; cGK; cGK 1; cGK1; cGKI; cGKI-alpha; cGKI-BETA; PKG; PRKG1; PRKG1B; PRKGR1A; PRKGR1B
UniProt ID: (Human) Q13976
Entrez Gene ID: (Human) 5592
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.