Novus Biologicals
Manufacturer Code:NBP171689
Catalog # NBP171689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A genomic peptide made to a C-terminal region of the human c11orf59/LAMTOR1 protein (within residues 50-250). [Swiss-Prot Q6IAA8] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 11 open reading frame 59 FLJ20625 LAMTOR1 Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1 late endosomal/lysosomal adaptor MAPK and MTOR activator 1 Lipid raft adaptor protein p18 p18 p27kip1 releasing factor from RhoA p27Kip1-releasing factor from RhoA p27RF-RhoC11orf59 PDRO PDROPdro Protein associated with DRMs and endosomes ragulator complex protein PDRO Ragulator1 RhoA activator C11orf59; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1; Lipid raft adaptor protein p18; p27kip1 releasing factor from RhoA; p27Kip1-releasing factor from RhoA; p27RF-Rho; Protein associated with DRMs and endosomes; Ragulator complex protein LAMTOR1; ragulator complex protein PDRO; RhoA activator C11orf59
Gene Aliases: C11orf59; LAMTOR1; p18; p27RF-Rho; PDRO; PP7157; Ragulator1
UniProt ID: (Human) Q6IAA8
Entrez Gene ID: (Human) 55004
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.