Invitrogen
Manufacturer Code:14611195
Catalog # 14-611-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 1RELAH5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: BCD541 C-Rel C-Rel protein C-Rel proto-oncogene protein gemin 1 gemin-1 oncogene REL avian reticuloendotheliosis Proto-oncogene c-Rel REL REL proto-oncogene NF-kB subunit reticuloendotheliosis oncogene SMA SMA1 SMA2 SMA3 SMA4 SMN SMN2 SMNT v-rel avian reticuloendotheliosis viral oncogene homolog v-rel reticuloendotheliosis viral oncogene homolog; C-Rel proto-oncogene protein; proto-oncogene c-Rel
Gene Aliases: c-Rel
Entrez Gene ID: (Mouse) 19696
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.