Novus Biologicals
Manufacturer Code:NBP237593
Catalog # NBP237593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10000000 |
Immunogen | Purified recombinant fragment of human c-Rel expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-Rel C-Rel proto-oncogene protein I-Rel oncogene REL avian reticuloendotheliosis proto-oncogene c-Rel v-rel avian reticuloendotheliosis viral oncogene homolog v-rel reticuloendotheliosis viral oncogene homolog (avian); oncogene REL, avian reticuloendotheliosis; Proto-oncogene c-Rel; v-rel avian reticuloendotheliosis viral oncogene homolog
Gene Aliases: C-Rel; REL
UniProt ID: (Human) Q04864
Entrez Gene ID: (Human) 5966
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.