Invitrogen
Manufacturer Code:PA568782
Catalog # PIPA568782
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse c-Rel |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-Rel proto-oncogene protein; oncogene REL, avian reticuloendotheliosis; Proto-oncogene c-Rel; REL v-rel reticuloendotheliosis viral oncogene homolog (avian) C-Rel C-Rel proto-oncogene protein oncogene REL avian reticuloendotheliosis v-rel avian reticuloendotheliosis viral oncogene homolog v-rel reticuloendotheliosis viral oncogene homolog; reticuloendotheliosis oncogene; v-rel avian reticuloendotheliosis viral oncogene homolog; v-rel reticuloendotheliosis viral oncogene homolog
Gene Aliases: C-Rel; REL
UniProt ID: (Human) Q04864
Entrez Gene ID: (Human) 5966, (Bovine) 509135, (Dog) 474606, (Horse) 100065217, (Rabbit) 100355271, (Mouse) 19696, (Rat) 305584, (Guinea pig) 100733398
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.