R&D Systems
Manufacturer Code:AF2699
Catalog # AF2699
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse c-Rel Met1-Ile588 Accession # NP_033070 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cRel C-Rel C-Rel proto-oncogene protein I-Rel oncogene REL avian reticuloendotheliosis proto-oncogene c-Rel REL v-rel avian reticuloendotheliosis viral oncogene homolog v-rel reticuloendotheliosis viral oncogene homolog (avian); oncogene REL, avian reticuloendotheliosis; Proto-oncogene c-Rel; v-rel avian reticuloendotheliosis viral oncogene homolog
Gene Aliases: C-Rel; REL
UniProt ID: (Human) Q04864
Entrez Gene ID: (Human) 5966
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.