Invitrogen
Manufacturer Code:MA116637
Catalog # AMPA6637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9E11 |
Immunogen | Synthetic peptide conjugated to KLH corresponding to residues A(408) E E Q K L I S E E D L(420) of human c-Myc. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc Cellular myelocytomatosis oncogene Myc protein Myc proto oncogene protein Myc2 Niard Nird v myc avian myelocytomatosis viral oncogene hom; c-Myc; c-myc proto-oncogene; Class E basic helix-loop-helix protein 39; Myc proto-oncogene protein; MYC proto-oncogene, bHLH transcription factor; myc-related translation/localization regulatory factor; Proto-oncogene c-Myc; Transcription factor p64; v-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene-like protein
Gene Aliases: AU016757; BHLHE39; c-Myc; MRTL; MYC; Myc2; MYCC; Niard; Nird
UniProt ID: (Chicken) P01109, (Human) P01106, (Mouse) P01108
Entrez Gene ID: (Chicken) 420332, (Human) 4609, (Mouse) 17869
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.