Invitrogen
Manufacturer Code:11885482
Catalog # 50-100-51
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBioclone 7 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AUTS9 HGFR RCCP2 c-Met; hepatocyte growth factor receptor; HGF receptor; HGF/SF receptor; met proto-oncogene tyrosine kinase; proto-oncogene c-Met; scatter factor receptor; SF receptor; tyrosine-protein kinase Met
Gene Aliases: AI838057; c-Met; HGF; HGFR; Par4
Entrez Gene ID: (Mouse) 17295
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.