Invitrogen
Manufacturer Code:14885437
Catalog # 14-885-437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBioclone 7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AI838057 AUTS9 bHLHe59 CG1705 CG1705 gene product from transcript CG1705-RB CG1705-PA CG1705-PB cMet c-met DFNB97 DmelCG1705 Dmel_CG1705 DmMet EC 2.7.10.1 Hepatocyte growth factor receptor HGF HGF receptor HGF receptor c-Met HGF/SF receptor Hgfr HGF-SF receptor juvenile hormone resistance Met met proto-oncogene met proto-oncogene (hepatocyte growth factor receptor) met proto-oncogene tyrosine kinase MET proto-oncogene receptor tyrosine kinase Met/Met1 methoprene tolerant Methoprene-tolerant Methoprene-tolerant protein Met-PA Met-PB Mett oncogene MET Par4 proto-oncogene c-Met RCCP2 resistance (1) juvenile H resistance to juvenile hormone Rst(1)JH sc MET Scatter factor receptor SF receptor soluble c met Tyrosine-protein kinase Met; hepatocyte growth factor receptor; HGF receptor; HGF/SF receptor; met proto-oncogene tyrosine kinase; proto-oncogene c-Met; scatter factor receptor; SF receptor; tyrosine-protein kinase Met
Gene Aliases: AI838057; c-Met; HGF; HGFR; Par4
Entrez Gene ID: (Mouse) 17295
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.