Invitrogen
Manufacturer Code:PA120184
Catalog # PIPA120184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit mouse hepatocyte growth factor receptor (HGFR). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AUTS9 HGFR RCCP2 c-Met; hepatocyte growth factor receptor; HGF receptor; HGF/SF receptor; met proto-oncogene tyrosine kinase; proto-oncogene c-Met; scatter factor receptor; SF receptor; tyrosine-protein kinase Met
Gene Aliases: AI838057; c-Met; HGF; HGFR; Par4
Entrez Gene ID: (Mouse) 17295
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.