Invitrogen
Manufacturer Code:MA529722
Catalog # PIMA529722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 17 |
Immunogen | Recombit Mouse KIT/c-KIT/CD117 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: belly-spot; C-Kit CD117 PBT SCFR mast/stem cell growth factor receptor Kit p145 c-kit piebald trait protein proto-oncogene c-Kit proto-oncogene tyrosine-protein kinase Kit soluble KIT variant 1 tyrosine-protein kinase Kit v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein ckit; c-kit proto-oncogene protein; CD117; dominant spotting; Dominant white spotting; Mast/stem cell growth factor receptor Kit; Proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; SCFR; spotted sterile male; Steel Factor Receptor; Tyrosine-protein kinase Kit
Gene Aliases: Bs; c-KIT; CD117; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; Kit; SCO1; SCO5; Sl; SOW3; Ssm; Tr-kit; W
UniProt ID: (Mouse) P05532
Entrez Gene ID: (Mouse) 16590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.