Invitrogen
Manufacturer Code:MA515055
Catalog # PIMA515055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | T.142.5 |
Immunogen | Synthetic peptide corresponding to the sequence of human c-Fos |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: activator protein 1; c-fos oncogene; c-fos protooncogene protein; cellular oncogene c-fos; Cellular oncogene fos; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS) G0/G1 switch regulatory protein 7 activator protein 1 cellular oncogene c-fos cellular oncogene fos proto-oncogene c-Fos v-fos FBJ murine osteosarcoma viral oncogene homolog; FBJ murine osteosarcoma viral oncogene homolog; FBJ osteosarcoma oncogene; Fos proto-oncogene, AP-1 trancription factor subunit; G0/G1 switch regulatory protein 7; Proto-oncogene c-Fos; proto-oncogene cFOS; Proto-oncogene protein c-fos; v-fos FBJ murine osteosarcoma viral oncogene homolog
Gene Aliases: AP-1; C-FOS; cFos; D12Rfj1; FOS; G0S7; I79_009428; p55
UniProt ID: (Bovine) O77628, (Human) P01100, (Pig) O97930, (Chinese hamster) Q9Z2Q8, (Mouse) P01101, (Rat) P12841
Entrez Gene ID: (Bovine) 280795, (Human) 2353, (Pig) 100144486, (Chinese hamster) 100689418, (Mouse) 14281, (Rat) 314322
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.