Invitrogen
Manufacturer Code:OSC00040W
Catalog # PIOSC00040W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from aa region 200-270 of rat c Fos protein conjugated to an immunogenic carrier protein has been used as the antigen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-fos oncogene; Cellular oncogene fos; FBJ murine osteosarcoma viral oncogene homolog; Proto-oncogene c-Fos; proto-oncogene cFOS; proto-oncogene protein c-fos; Proto-oncogene protein cFOS c-Fos; v-fos FBJ murine osteosarcoma viral oncogene homolog
Gene Aliases: c-fos; cFos; D12Rfj1; Fos
UniProt ID: (Mouse) P01101, (Rat) P12841
Entrez Gene ID: (Mouse) 14281, (Rat) 314322
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.