Novus Biologicals
Manufacturer Code:NB600904
Catalog # NB600904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (Human) (C terminal). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activator protein 1; activator protein 1 AP-1 cellular oncogene c-fos Cellular oncogene fos c-fos FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS) FBJ murine osteosarcoma viral oncogene homolog G0/G1 switch regulatory protein 7 G0S7 proto-oncogene c-Fos v-fos FBJ murine osteosarcoma viral oncogene homolog; cellular oncogene c-fos; Cellular oncogene fos; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; Fos proto-oncogene, AP-1 trancription factor subunit; G0/G1 switch regulatory protein 7; Proto-oncogene c-Fos
Gene Aliases: AP-1; C-FOS; FOS; G0S7; p55
UniProt ID: (Human) P01100
Entrez Gene ID: (Human) 2353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.