Invitrogen
Manufacturer Code:MA514398
Catalog # PIMA514398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8E9 |
Immunogen | Recombit Abl protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine-protein kinase 1; bcr/c-abl oncogene protein; bcr/c-abl oncogene protein proto-oncogene c-Abl proto-oncogene tyrosine-protein kinase ABL1 tyrosine-protein kinase ABL1 v-abl Abelson murine leukemia viral oncogene homolog 1; c-abl oncogene 1, receptor tyrosine kinase; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; Tyrosine-protein kinase ABL1; v-abl Abelson murine leukemia viral oncogene homolog 1
Gene Aliases: ABL; ABL1; bcr/abl; c-ABL; c-ABL1; JTK7; p150; v-abl
UniProt ID: (Human) P00519
Entrez Gene ID: (Human) 25
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.