Novus Biologicals
Manufacturer Code:NBP299737G
Catalog # NB086173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant c-Abl (NP_009297.2 Pro137-Ser554). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson murine leukemia viral oncogene homolog 1; Abelson murine leukemia viral oncogene homolog 1 ABL c-ABL c-abl oncogene 1 non-receptor tyrosine kinase c-abl oncogene 1 receptor tyrosine kinase EC 2.7.10 EC 2.7.10.2 JTK7bcr/abl p150bcr/c-abl oncogene protein Proto-oncogene c-Abl proto-oncogene tyrosine-protein kinase ABL1 tyrosine-protein kinase ABL1 v-abl v-abl Abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine-protein kinase 1; bcr/c-abl oncogene protein; c-abl oncogene 1, receptor tyrosine kinase; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; Tyrosine-protein kinase ABL1; v-abl Abelson murine leukemia viral oncogene homolog 1
Gene Aliases: ABL; ABL1; bcr/abl; c-ABL; c-ABL1; JTK7; p150; v-abl
UniProt ID: (Human) P00519
Entrez Gene ID: (Human) 25
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.