R&D Systems
Manufacturer Code:AF5414
Catalog # AF5414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human c-Abl Ala941-Val1140 Accession # NP_009297 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson murine leukemia viral oncogene homolog 1; Abelson murine leukemia viral oncogene homolog 1 ABL ABL1 bcr/abl BCR-ABL1 cAbl c-ABL c-abl oncogene 1 non-receptor tyrosine kinase c-abl oncogene 1 receptor tyrosine kinase EC 2.7.10 EC 2.7.10.2 JTK7 JTK7bcr/abl p150bcr/c-abl oncogene protein Proto-oncogene c-Abl proto-oncogene tyrosine-protein kinase ABL1 tyrosine-protein kinase ABL1 v-abl v-abl Abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine-protein kinase 1; bcr/c-abl oncogene protein; c-abl oncogene 1, receptor tyrosine kinase; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; Tyrosine-protein kinase ABL1; v-abl Abelson murine leukemia viral oncogene homolog 1
Gene Aliases: ABL; ABL1; bcr/abl; c-ABL; c-ABL1; JTK7; p150; v-abl
UniProt ID: (Human) P00519
Entrez Gene ID: (Human) 25
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.