Abnova
Manufacturer Code:H00011149M02
Catalog # 89-014-277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F7 |
Immunogen | BVES (NP_009004 262 a.a. ~ 360 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant BVES.
For Research Use Only
Protein Aliases: Blood vessel epicardial substance; HBVES; MGC42413; POP1; POPDC1; Popeye domain-containing protein 1; Popeye protein 1
Gene Aliases: BVES; HBVES; LGMD2X; POP1; POPDC1
UniProt ID: (Human) Q8NE79
Entrez Gene ID: (Human) 11149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.