Invitrogen
Manufacturer Code:MA516308
Catalog # PIMA516308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BT7R |
Immunogen | KLH conjugated peptide of the N-terminus of Beta-Tubulin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta 5-tubulin; beta 5-tubulin beta Ib tubulin beta-4 tubulin tubulin beta chain tubulin beta polypeptide tubulin beta-1 chain tubulin beta-5 chain tubulin beta polypeptide; beta Ib tubulin; beta-tubulin; Beta-tubulin class-VI; c(beta)7 tubulin; class I beta-tubulin; Tubulin beta 4'; Tubulin beta chain; Tubulin beta-1 chain; Tubulin beta-5 chain; Tubulin beta-6 chain; Tubulin beta-7 chain; tubulin, beta 1 class VI; tubulin, beta 2A; tubulin, beta 2A class IIa; tubulin, beta 3; tubulin, beta class I; tubulin, beta polypeptide; unnamed protein product
Gene Aliases: 2810484G07Rik; CDCBM6; CSCSC1; M(beta)1; M40; OK/SW-cl.56; TUBB; TUBB1; TUBB5
UniProt ID: (Human) P07437, (Chicken) P09244, (Chicken) P09207, (Human) Q9H4B7, (Mouse) A2AQ07
Entrez Gene ID: (Dog) 474830, (Human) 203068, (Rabbit) 100358689, (Chicken) 396254, (Chicken) 396427, (Human) 81027, (Rabbit) 100347858, (Mouse) 545486, (Rat) 679312
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.