Novus Biologicals
Manufacturer Code:NBP271401AF700
Catalog # N271401A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D5 |
Immunogen | Recombinant protein fragment corresponding to amino acids 52-354 of human beta-TrCP1/BTRC (NP_378663) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: beta-transducin repeat containing betaTrCP BETA-TRCP beta-TrCP1 bTrCP bTrCP1 E3RSIkappaB F-box and WD repeats protein beta-TrCP F-box/WD repeat-containing protein 1A FBW1A FBXW1 FBXW1ABTRCP Fwd1 MGC4643 pIkappaBalpha-E3 receptor subunit; beta-TrCP1; E3RSIkappaB; Epididymis tissue protein Li 2a; F-box and WD repeats protein beta-TrCP; F-box and WD-repeat protein 1B; F-box/WD repeat-containing protein 1A; pIkappaBalpha-E3 receptor subunit
Gene Aliases: BETA-TRCP; betaTrCP; BTRC; BTRCP; bTrCP1; FBW1A; FBXW1; FBXW1A; FWD1
UniProt ID: (Human) Q9Y297
Entrez Gene ID: (Human) 8945
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.