R&D Systems
Manufacturer Code:MAB556
Catalog # MAB556
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 884530 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant rat beta-NGF Met1-Gly241 Accession # P25427 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
Protein Aliases: Beta nerve growth factor precursor beta-nerve growth factor betaNGF Beta-NGF HSAN5 MGC161426 nerve growth factor (beta polypeptide) nerve growth factor beta subunit NGFB NGFBMGC161428; Beta-nerve growth factor; Beta-NGF; nerve growth factor (beta polypeptide); nerve growth factor, beta subunit
Gene Aliases: Beta-NGF; HSAN5; NGF; NGFB
UniProt ID: (Human) P01138
Entrez Gene ID: (Human) 4803
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.