Novus Biologicals
Manufacturer Code:NB200117SS
Catalog # NB200117SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of Defensin beta 3 (between amino acids 10-50) [UniProt P81534] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BD-3; BD3 BD-3 beta-defensin 103 Beta-defensin 3 DEFB103 DEFB-3 DEFB3DEFB103A Defensin beta 103 defensin beta 103B Defensin-like protein hBD-3 HBD3HBP3; Beta-defensin 103; Beta-defensin 3; Defensin, beta 103; defensin, beta 103B; Defensin-like protein
Gene Aliases: BD-3; BD3; DEFB-3; DEFB103; DEFB103A; DEFB103B; DEFB3; HBD-3; HBD3; HBP-3; HBP3
UniProt ID: (Human) P81534
Entrez Gene ID: (Human) 55894
Molecular Function:
antibacterial response protein
defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.