Invitrogen
Manufacturer Code:PA5113860
Catalog # PIPA5113860
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ARRB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI326910 ARB2 ARR2 ARRB2 arrestin arrestin 3 arrestin beta 2 Arrestin beta-2 arrestin beta 2 arrestin-3 AW122872 BARR2 BARRES beta arr2 beta-arrestin 2 beta-arrestin2 Beta-arrestin-2 BOS_18747 DKFZp686L0365 HGNC:712 Non-visual arrestin-3 SAG S-Arrestin; arrestin 3; Arrestin beta-2; arrestin, beta 2; beta arr2; Beta-arrestin-2; Non-visual arrestin-3
Gene Aliases: AI326910; ARB2; ARR2; ARRB2; AW122872; BARR2
UniProt ID: (Human) P32121, (Mouse) Q91YI4
Entrez Gene ID: (Human) 409, (Mouse) 216869
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.