Invitrogen
Manufacturer Code:MA537831
Catalog # PIMA537831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2370 |
Immunogen | Recombinant protein of human ARRB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 1200006I17Rik ARB1 ARR1 ARRB1 ARRB1 protein arrdc1 arrestin arrestin 2 arrestin beta 1 arrestin beta-1 arrestin beta 1 arrestin-2 AW208571 BARRES beta-arrestin beta-arrestin1 beta-arrestin-1 G430100A01Rik Non-visual arrestin-2; arrestin 2; Arrestin beta-1; arrestin, beta 1; Beta-arrestin-1; beta-arrestin1; Non-visual arrestin-2
Gene Aliases: 1200006I17Rik; ARB1; ARR1; ARRB1; AW208571; G430100A01Rik
UniProt ID: (Human) P49407, (Mouse) Q8BWG8
Entrez Gene ID: (Human) 408, (Mouse) 109689
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.