R&D Systems
Manufacturer Code:MAB45601SP
Catalog # MAB45601SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 425817 |
Immunogen | <EM>E. coli</EM>-derived recombinant human beta-Arrestin 1 Met1-Arg418 Accession # P49407 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ARB1 ARR1 ARRB1 arrestin 2 Arrestin beta-1 arrestin beta 1 betaArrestin 1 beta-arrestin-1; arrestin 2; Arrestin beta-1; arrestin, beta 1; Beta-arrestin-1; Non-visual arrestin-2
Gene Aliases: ARB1; ARR1; ARRB1
UniProt ID: (Human) P49407
Entrez Gene ID: (Human) 408
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.