Invitrogen
Manufacturer Code:PA519582
Catalog # PIPA519582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 400 to the C-terminus of Mouse beta Arrestin 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARB1 ARR1 arrestin 2 beta-arrestin-1; Arrestin beta-1; Beta-arrestin-1; beta-arrestin1
Gene Aliases: 1200006I17Rik; Arrb1; AW208571; BARRES; G430100A01Rik
UniProt ID: (Rat) P29066, (Mouse) Q8BWG8
Entrez Gene ID: (Rat) 25387, (Mouse) 109689
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.