Proteintech
Manufacturer Code:255241AP150UL
Catalog # 50-173-4947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | beta Amyloid (C-Terminal) Fusion Protein Ag22408 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A4 AAA ABETA ABPP AD1 AICD 50 AICD 57 AICD 59 AID(50) AID(57) AID(59) Amyloid beta A4 protein amyloid P APP APPI beta Amyloid CTFgamma CVAP Gamma CTF(50) C31 PN II PN2 PreA4 Protease nexin II S APP alpha Soluble APP beta; Abeta40; Abeta42; ABPP; AG; AICD-50; AICD-57; AICD-59; AID(50); AID(57); AID(59); Alpha-CTF; Alpha-secretase C-terminal fragment; Alzheimer disease amyloid A4 protein homolog; Alzheimer disease amyloid protein; amyloid beta (A4) precursor protein; amyloid beta A4 protein; Amyloid intracellular domain 50; Amyloid intracellular domain 57; Amyloid intracellular domain 59; Amyloid precursor protein; Amyloid-beta A4 protein; Amyloid-beta precursor protein; Amyloid-beta protein 40; Amyloid-beta protein 42; Amyloidogenic glycoprotein; APP; APP-C57; APP-C59; APP-C99; APPI; appican; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); beta-amyloid precursor protein; Beta-APP40; Beta-APP42; Beta-CTF; Beta-secretase C-terminal fragment; C31; C80; C83; C99; Cerebral vascular amyloid peptide; CVAP; Gamma-CTF(50); Gamma-CTF(57); Gamma-CTF(59); Gamma-secretase C-terminal fragment 50; Gamma-secretase C-terminal fragment 57; Gamma-secretase C-terminal fragment 59; N-APP; P3(40); P3(42); peptidase nexin-II; PN-II; PreA4; protease nexin II; Protease nexin-II; S-APP-alpha; S-APP-beta; Soluble APP-alpha; Soluble APP-beta; testicular tissue protein Li 2
Gene Aliases: A4; AAA; ABETA; ABPP; AD1; Adap; Ag; APP; APPI; betaApp; CTFgamma; CVAP; E030013M08Rik; PN-II; PN2
UniProt ID: (Human) P05067, (Mouse) P12023
Entrez Gene ID: (Human) 351, (Mouse) 11820
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.