Novus Biologicals
Manufacturer Code:NB600501SS
Catalog # NB600501SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AC-15 |
Immunogen | This beta-Actin Antibody (AC-15) was made to a slightly modified Beta-cytoplasmic actin N-terminal peptide Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys conjugated to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACTB actin beta actin cytoplasmic 1 anti-beta actin B-Actin Beta Actin beta actin (ac-15) beta cytoskeletal actin Beta-actin beta-actin ac 15 beta-actin ac15 beta-actin ac-15 BRWS1 I(2)-Actin PS1TP5-binding protein 1 PS1TP5BP1; actin, beta; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; beta cytoskeletal actin; Beta-actin; PS1TP5-binding protein 1
Gene Aliases: ACTB; BRWS1; PS1TP5BP1
UniProt ID: (Human) P60709
Entrez Gene ID: (Human) 60
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.