Invitrogen
Manufacturer Code:PA585271
Catalog # PIPA585271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human beta Actin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin, beta; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; beta actin; beta cytoskeletal actin; Beta-actin; Cytoplasmic beta-actin; cytoskeletal beta actin; PS1TP5-binding protein 1; PS1TP5BP1 antibody ACTB antibody actin cytoplasmic 1 antibody beta cytoskeletal actin antibody PS1TP5-binding protein 1 antibody actin beta antibody
Gene Aliases: ACTB; Actx; beta-actin; BRWS1; E430023M04Rik; PS1TP5BP1; XELAEV_18045052mg
UniProt ID: (Horse) P60708, (Human) P60709, (Pig) Q6QAQ1, (Dog) O18840, (Rat) P60711, (Mouse) P60710, (Xenopus) O93400
Entrez Gene ID: (Horse) 100033878, (Human) 60, (Pig) 414396, (Dog) 403580, (Rat) 81822, (Mouse) 11461, (Xenopus) 398459
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.