Novus Biologicals
Manufacturer Code:NB600532SS
Catalog # NB600532SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This beta-Actin Antibody maps to a region corresponding to the N-terminus of human Beta Actin. [UniProt# P60709] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACTB actin beta actin cytoplasmic 1 B-Actin Beta Actin beta cytoskeletal actin Beta-actin BRWS1 I(2)-Actin PS1TP5-binding protein 1 PS1TP5BP1; actin, beta; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; beta cytoskeletal actin; Beta-actin; PS1TP5-binding protein 1
Gene Aliases: ACTB; BRWS1; PS1TP5BP1
UniProt ID: (Human) P60709
Entrez Gene ID: (Human) 60
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.