R&D Systems
Manufacturer Code:FAB9419T100UG
Catalog # FAB9419T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 493324 |
Immunogen | Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3 Met1-Ser408 Accession # P13945 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: ADRB3 ADRB3R adrenergic beta-3- receptor B3AR beta-3 adrenergic receptor beta3 AdrenergicR beta-3 AdrenergicR Beta-3 adrenoceptor Beta-3 adrenoreceptor BETA3AR FLJ99960; adrenergic, beta-3-, receptor; Beta-3 adrenergic receptor; Beta-3 adrenoceptor; Beta-3 adrenoreceptor
Gene Aliases: ADRB3; ADRB3R; B3AR; BETA3AR
UniProt ID: (Human) P13945
Entrez Gene ID: (Human) 155
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.