Invitrogen
Manufacturer Code:53451082
Catalog # 50-112-4599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G10-TB3 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: betaIII-tubulin; class III beta-tubulin; neuron specific beta III Tubulin; Neuron-specific class III beta-tubulin; Tubulin beta-3 chain; Tubulin beta-4 chain; Tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4
Gene Aliases: 3200002H15Rik; beta-4; CDCBM; CDCBM1; CFEOM3; CFEOM3A; FEOM3; M(beta)3; M(beta)6; TUBB3; TUBB4
UniProt ID: (Bovine) Q2T9S0, (Human) Q13509, (Mouse) Q9ERD7, (Rat) Q4QRB4
Entrez Gene ID: (Bovine) 768070, (Human) 10381, (Mouse) 22152, (Rat) 246118
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.