Invitrogen
Manufacturer Code:MA515541
Catalog # PIMA515541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E9 |
Immunogen | Purified recombit fragment of human TUBB3 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: class III beta-tubulin; tubulin beta 3 MC1R TUBB4 beta-4 TUBB3; Tubulin beta-3 chain; Tubulin beta-4 chain; Tubulin beta-III; tubulin, beta 3 class III
Gene Aliases: beta-4; CDCBM; CDCBM1; CFEOM3; CFEOM3A; FEOM3; TUBB3; TUBB4
UniProt ID: (Human) Q13509
Entrez Gene ID: (Human) 10381
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.