Invitrogen
Manufacturer Code:PA581099
Catalog # PIPA581099
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the Human TUBB3/beta III Tubulin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: class III beta-tubulin; neuron specific beta III Tubulin; Tubulin beta-3 chain; Tubulin beta-4 chain; Tubulin beta-III; tubulin, beta 3 class III
Gene Aliases: beta-4; CDCBM; CDCBM1; CFEOM3; CFEOM3A; FEOM3; TUBB3; TUBB4
UniProt ID: (Human) Q13509
Entrez Gene ID: (Human) 10381
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.