Invitrogen
Manufacturer Code:PA5114219
Catalog # PIPA5114219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: LHWYTGEGMD EMEFTEAESN MNDLVSEYQQ YQDATAEEEG EMYEDDEEES |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3200002H15Rik beta tubulin 3 beta-4 beta-4-tubulin betaIII-tubulin Beta-tubulin class-III BOS_17166 CDCBM CDCBM1 CFEOM3 CFEOM3A class III beta-tubulin FEOM3 I79_000194 LOC100341581 M(beta)3 M(beta)6 MC1R Neuron-specific class III beta-tubulin TUBB TUBB3 TUBB4 tubulin beta tubulin beta 3 class III Tubulin beta-3 chain Tubulin beta-4 chain Tubulin beta-III tubulin beta 3 class III tubulin beta 3 tubulin beta 4 tubulin beta 4-like; class III beta-tubulin; Tubulin beta-3 chain; Tubulin beta-4 chain; Tubulin beta-III; tubulin, beta 3 class III
Gene Aliases: beta-4; CDCBM; CDCBM1; CFEOM3; CFEOM3A; FEOM3; TUBB3; TUBB4
UniProt ID: (Human) Q13509
Entrez Gene ID: (Human) 10381
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.