Invitrogen
Manufacturer Code:PA5114902
Catalog # PIPA5114902
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human beta Defensin 3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BD-3; BD3 BD-3 beta defensin 3 beta defensin-3 beta-defensin 103 beta-defensin 3 DEFB103 DEFB103A DEFB103B DEFB3 DEFB-3 defensin beta 103A defensin beta 103 defensin beta 103A defensin beta 3 Defensin-like protein HBD3 hBD-3 HBP3 HBP-3; beta defensin 3; beta defensin-3; Beta-defensin 103; Beta-defensin 3; Defensin, beta 103; defensin, beta 103A; defensin, beta 3; Defensin-like protein
Gene Aliases: BD-3; BD3; DEFB-3; DEFB103; DEFB103A; DEFB103B; DEFB3; hBD-3; HBD3; HBP-3; HBP3
UniProt ID: (Human) P81534
Entrez Gene ID: (Human) 414325
Molecular Function:
antibacterial response protein
defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.