Invitrogen
Manufacturer Code:PA577284
Catalog # PIPA577284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide DAEAQESH*SNPRC corresponding to amino acid residues 180-192 of mouse beta3 -Adrenoceptor with replacement of cysteine 186 (C186 ) with serine (*S) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenergic receptor, beta 3; adrenergic, beta-3-, receptor; beta 3-AR; Beta-3 adrenergic receptor; Beta-3 adrenoceptor; Beta-3 adrenoreceptor; Beta-3 AR; beta3-adrenergic receptor; beta3-Adrenoreceptor ADRB3
Gene Aliases: ADRB; Adrb-3; Adrb3; Adrb3r; B3bar
UniProt ID: (Mouse) P25962, (Rat) P26255
Entrez Gene ID: (Mouse) 11556, (Rat) 25645
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.