Invitrogen
Manufacturer Code:PA533336
Catalog # PIPA533336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from C-terminus of human ADRB3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRB3 Adrenoceptor beta 3 ADRB3R Adrenaline beta 3 receptor Adrenergic beta-3- receptor Beta-3 adrenergic receptor B3AR Beta-3 adrenoceptor Beta3-AR Beta-3 adrenoreceptor BETA3AR; adrenergic, beta-3-, receptor; Beta-3 adrenergic receptor; Beta-3 adrenoceptor; Beta-3 adrenoreceptor
Gene Aliases: ADRB3; ADRB3R; B3AR; BETA3AR
UniProt ID: (Human) P13945
Entrez Gene ID: (Human) 155
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.